  • Search Results
  • Cytochrome c oxidase inhibition by calcium at physiological ionic composition of the medium: Implications for physiological significance of the effect
Cytochrome c oxidase inhibition by calcium at physiological ionic composition of the medium: Implications for physiological significance of the effect
Biochimica et Biophysica Acta - Bioenergetics (2017)
Vygodina T, et al.