  • Search Results
  • Effect of choline chloride on the lipid content and composition in the leaves of principal magnetically-oriented radish types
Effect of choline chloride on the lipid content and composition in the leaves of principal magnetically-oriented radish types
Russian Journal of Plant Physiology (2004)
Novitskaya GV, et al.