  • Search Results
  • Bioconjugation of magnetic iron oxide nanoparticles and cross-linked enzyme aggregates: characterization and comparison of CLEAs and magnetic CLEAs
Bioconjugation of magnetic iron oxide nanoparticles and cross-linked enzyme aggregates: characterization and comparison of CLEAs and magnetic CLEAs
Current Nanoscience (2017)
Salgin S and Salgin U