  • Search Results
  • Identification and pharmacological characterization of a novel inhibitor of autotaxin in rodent models of joint pain
Identification and pharmacological characterization of a novel inhibitor of autotaxin in rodent models of joint pain
Osteoarthritis and Cartilage (2017)
Thirunavukkarasu K, et al.