  • Search Results
  • Pathophysiological roles and clinical importance of biomarkers in acute coronary syndrome
Pathophysiological roles and clinical importance of biomarkers in acute coronary syndrome
Advances in Clinical Chemistry (2013)
Chan CP and Rainer TH