  • Search Results
  • Magnesium sulphate and other anticonvulsants for women with pre-eclampsia
Magnesium sulphate and other anticonvulsants for women with pre-eclampsia
Cochrane database of systematic reviews (Online) (2010)
Duley L, et al.