  • Search Results
  • ELISA to evaluate plasma anti-asparaginase IgG concentrations in patients with acute lymphoblastic leukemia
ELISA to evaluate plasma anti-asparaginase IgG concentrations in patients with acute lymphoblastic leukemia
Journal of Immunological Methods (2000)
Wang B, et al.