  • Search Results
  • Effects of treatment with fluoride on bone mineral density and fracture risk-a meta-analysis
Effects of treatment with fluoride on bone mineral density and fracture risk-a meta-analysis
Osteoporosis International : A Journal Established as Result of Cooperation Between the European Foundation for Osteoporosis and the National Osteoporosis Foundation of the USA (2008)
Vestergaard P, et al.