  • Search Results
  • Fluoride-induced oxidative stress and apoptosis are involved in the reducing of oocytes development potential in mice
Fluoride-induced oxidative stress and apoptosis are involved in the reducing of oocytes development potential in mice
Chemosphere (2017)
Wang HW, et al.