MilliporeSigma
Gut-liver on a chip toward an in vitro model of hepatic steatosis.
Biotechnology and bioengineering (2018-07-08)
Seung Yeon Lee, Jong Hwan Sung
ABSTRACT
Hepatic steatosis is a process of abnormal lipid deposition within the liver cells, often caused by excessive alcohol uptake or obesity. A conventional in vitro model for hepatic steatosis uses a liver cell culture, treated with fatty acids and measures accumulation of lipids within the cells. This model does not recapitulate the complex process of absorption and metabolism of digestive lipids. Here, we introduce a gut-liver chip, which mimics the gut absorption and hepatic metabolism in a microfluidic chip. Absorption of fatty acids through gut layer and subsequent deposition within liver cells was demonstrated. Tumor necrosis factor-α, butyrate, and α-lipoic acid were chosen as model molecules that can affect hepatic steatosis via different mechanisms, and their effects were evaluated. Our results suggest that the gut-liver chip can mimic the absorption and accumulation of fatty acids in the gut and the liver.
Sigma-Aldrich
Hanks′ Balanced Salt solution, Modified, with sodium bicarbonate, without phenol red, calcium chloride and magnesium sulfate, liquid, sterile-filtered, suitable for cell culture
Sigma-Aldrich
Fluorescein isothiocyanate–dextran, average mol wt 4,000, (FITC:Glucose = 1:250)