  • Search Results
  • MID1 and MID2 are required for Xenopus neural tube closure through the regulation of microtubule organization.
MID1 and MID2 are required for Xenopus neural tube closure through the regulation of microtubule organization.
Development (2010)
Suzuki M, et al.