  • Search Results
  • Analysis of gene function in somatic mammalian cells using small interfering RNAs.
Analysis of gene function in somatic mammalian cells using small interfering RNAs.
Methods (2002)
Elbashir SM, et al.