  • Search Results
  • TRAIL induces apoptosis in triple-negative breast cancer cells with a mesenchymal phenotype.
TRAIL induces apoptosis in triple-negative breast cancer cells with a mesenchymal phenotype.
Breast Cancer Research and Treatment (2009)
Rahman M, et al.