MilliporeSigma
  • Defective endothelial cell migration in the absence of Cdc42 leads to capillary-venous malformations.
Defective endothelial cell migration in the absence of Cdc42 leads to capillary-venous malformations.
Development (Cambridge, England) (2018-06-02)
Bàrbara Laviña, Marco Castro, Colin Niaudet, Bert Cruys, Alberto Álvarez-Aznar, Peter Carmeliet, Katie Bentley, Cord Brakebusch, Christer Betsholtz, Konstantin Gaengel
ABSTRACT
Formation and homeostasis of the vascular system requires several coordinated cellular functions, but their precise interplay during development and their relative importance for vascular pathologies remain poorly understood. Here, we investigated the endothelial functions regulated by Cdc42 and their in vivo relevance during angiogenic sprouting and vascular morphogenesis in the postnatal mouse retina. We found that Cdc42 is required for endothelial tip cell selection, directed cell migration and filopodia formation, but dispensable for cell proliferation or apoptosis. Although the loss of Cdc42 seems generally compatible with apical-basal polarization and lumen formation in retinal blood vessels, it leads to defective endothelial axial polarization and to the formation of severe vascular malformations in capillaries and veins. Tracking of Cdc42-depleted endothelial cells in mosaic retinas suggests that these capillary-venous malformations arise as a consequence of defective cell migration, when endothelial cells that proliferate at normal rates are unable to re-distribute within the vascular network.
Sigma-Aldrich
Collagenase from Clostridium histolyticum, suitable for release of physiologically active rat epididymal adipocytes, Type II, 0.5-5.0 FALGPA units/mg solid, ≥125 CDU/mg solid
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Partitioning-defective 3 Antibody, Upstate®, from rabbit
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds