  • Search Results
  • Alpha-synuclein delays mitophagy and targeting Miro rescues neuron loss in Parkinson's models.
Alpha-synuclein delays mitophagy and targeting Miro rescues neuron loss in Parkinson's models.
Acta neuropathologica (2018-06-21)
Atossa Shaltouki, Chung-Han Hsieh, Min Joo Kim, Xinnan Wang
ABSTRACT
Alpha-synuclein is a component of Lewy bodies, the pathological hallmark of Parkinson's disease (PD), and is also mutated in familial PD. Here, by extensively analyzing PD patient brains and neurons, and fly models, we show that alpha-synuclein accumulation results in upregulation of Miro protein levels. Miro is a motor/adaptor on the outer mitochondrial membrane that mediates mitochondrial motility, and is removed from damaged mitochondria to facilitate mitochondrial clearance via mitophagy. PD patient neurons abnormally accumulate Miro on the mitochondrial surface leading to delayed mitophagy. Partial reduction of Miro rescues mitophagy phenotypes and neurodegeneration in human neurons and flies. Upregulation of Miro by alpha-synuclein requires an interaction via the N-terminus of alpha-synuclein. Our results highlight the importance of mitochondria-associated alpha-synuclein in human disease, and present Miro as a novel therapeutic target.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Sigma-Aldrich
Triton X-100, laboratory grade
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Tetramethylrhodamine methyl ester perchlorate, ≥95%
Sigma-Aldrich
Monoclonal Anti-α-Fetoprotein (AFP) antibody produced in mouse, ascites fluid, clone C3
Sigma-Aldrich
Anti-β-Tubulin III antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Tyrosine Hydroxylase Antibody, clone LNC1, ascites fluid, clone LNC1, Chemicon®
Sigma-Aldrich
Anti-SOX-2 Antibody, clone 6F1.2, Chemicon®, from mouse
Sigma-Aldrich
Anti-RHOT1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Sigma-Aldrich
Saponin, for molecular biology, used as non-ionic surfactant
Sigma-Aldrich
Monoclonal Anti-Actin, α-Smooth Muscle, clone 1A4, ascites fluid