  • Search Results
  • The size, shape, and hydration of nonionic surfactant micelles. Triton X-100.
The size, shape, and hydration of nonionic surfactant micelles. Triton X-100.
The Journal of Physical Chemistry (1977)
Robson RJ and Dennis EA