  • Search Results
  • Cytotoxicity of bilirubin for human fibroblasts and rat astrocytes in culture. Effect of the ratio of bilirubin to serum albumin.
Cytotoxicity of bilirubin for human fibroblasts and rat astrocytes in culture. Effect of the ratio of bilirubin to serum albumin.
Clinica Chimica Acta; International Journal of Clinical Chemistry (1996)
Chuniaud L, et al.