  • Search Results
  • Cardiolipin Synthesis in Brown and Beige Fat Mitochondria Is Essential for Systemic Energy Homeostasis.
Cardiolipin Synthesis in Brown and Beige Fat Mitochondria Is Essential for Systemic Energy Homeostasis.
Cell metabolism (2018-06-05)
Elahu G Sustarsic, Tao Ma, Matthew D Lynes, Michael Larsen, Iuliia Karavaeva, Jesper F Havelund, Carsten H Nielsen, Mark P Jedrychowski, Marta Moreno-Torres, Morten Lundh, Kaja Plucinska, Naja Z Jespersen, Trisha J Grevengoed, Barbara Kramar, Julia Peics, Jakob B Hansen, Farnaz Shamsi, Isabel Forss, Ditte Neess, Susanne Keipert, Jianing Wang, Katharina Stohlmann, Ivan Brandslund, Cramer Christensen, Marit E Jørgensen, Allan Linneberg, Oluf Pedersen, Michael A Kiebish, Klaus Qvortrup, Xianlin Han, Bente Klarlund Pedersen, Martin Jastroch, Susanne Mandrup, Andreas Kjær, Steven P Gygi, Torben Hansen, Matthew P Gillum, Niels Grarup, Brice Emanuelli, Søren Nielsen, Camilla Scheele, Yu-Hua Tseng, Nils J Færgeman, Zachary Gerhart-Hines
ABSTRACT
Activation of energy expenditure in thermogenic fat is a promising strategy to improve metabolic health, yet the dynamic processes that evoke this response are poorly understood. Here we show that synthesis of the mitochondrial phospholipid cardiolipin is indispensable for stimulating and sustaining thermogenic fat function. Cardiolipin biosynthesis is robustly induced in brown and beige adipose upon cold exposure. Mimicking this response through overexpression of cardiolipin synthase (Crls1) enhances energy consumption in mouse and human adipocytes. Crls1 deficiency in thermogenic adipocytes diminishes inducible mitochondrial uncoupling and elicits a nuclear transcriptional response through endoplasmic reticulum stress-mediated retrograde communication. Cardiolipin depletion in brown and beige fat abolishes adipose thermogenesis and glucose uptake, which renders animals insulin resistant. We further identify a rare human CRLS1 variant associated with insulin resistance and show that adipose CRLS1 levels positively correlate with insulin sensitivity. Thus, adipose cardiolipin has a powerful impact on organismal energy homeostasis through thermogenic fat bioenergetics.
Sigma-Aldrich
Antimycin A from Streptomyces sp.
Supelco
Docosanoic acid tryptamide, analytical standard
Sigma-Aldrich
Corn oil, delivery vehicle for fat-soluble compounds
Sigma-Aldrich
Rosiglitazone, ≥98% (HPLC)
Sigma-Aldrich
Insulin solution human, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Carbonyl cyanide 4-(trifluoromethoxy)phenylhydrazone, ≥98% (TLC), powder
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, fatty acid free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
GenElute Mammalian Genomic DNA Miniprep Kits, sufficient for 70 purifications
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium, Without glucose, L-glutamine, phenol red, sodium pyruvate and sodium bicarbonate, powder, suitable for cell culture