  • Search Results
  • Determination of trace amounts of phenol in river waters by gas chromatography
Determination of trace amounts of phenol in river waters by gas chromatography
Analyst (1982)
Rennie.JP