MilliporeSigma
  • P-glycoprotein inhibition as a therapeutic approach for overcoming multidrug resistance in cancer: current status and future perspectives.
P-glycoprotein inhibition as a therapeutic approach for overcoming multidrug resistance in cancer: current status and future perspectives.
Current cancer drug targets (2013-02-02)
Ziyad Binkhathlan, Afsaneh Lavasanifar
ABSTRACT
One of the major causes of failure in cancer chemotherapy is multidrug resistance (MDR), where cancer cells simultaneously become resistant to different anticancer drugs. Over-expression of membrane efflux pumps like P-glycoprotein (P-gp) that recognizes different chemotherapeutic agents and transports them out of the cell, plays a major role in MDR. The shortcoming of P-gp inhibitors in clinic has been attributed to their non-specific action on P-gp and/or non-selective distribution to non-target organs that leads to intolerable side effects by the P-gp inhibitor at doses required for P-gp inhibition upon systemic administration. Another major issue is the reduced elimination of P-gp substrates (e.g. anticancer drugs) and intolerable toxicities by anticancer drugs when co-administered with P-gp inhibitors. To overcome these shortcomings, new generation of P-gp inhibitors with improved specificity for P-gp have been developed. More recently, attention has been paid to the use of drug delivery systems primarily to restrict P-gp inhibition to tumor and reduce the non-selective inhibition of P-gp in non-target organs. This review will provide an overview and update on the status of P-gp inhibition approaches and the role of drug delivery systems in overcoming P-gp mediated MDR.