  • Search Results
  • Effects of processing on proximate and fatty acid compositions of six commercial sea cucumber species of Sri Lanka.
Effects of processing on proximate and fatty acid compositions of six commercial sea cucumber species of Sri Lanka.
Journal of food science and technology (2018-04-19)
G Nishanthan, P A D A Kumara, M D S T de Croos, D V P Prasada, D C T Dissanayake
ABSTRACT
Processing and its impacts on proximate composition and fatty acid profile of six sea cucumber species; Bohadschia marmorata, Stichopus chloronotus, Holothuria spinifera, Thelenota anax, Holothuria scabra and Bohadschia sp. 1 collected from the northwest coast of Sri Lanka were analyzed. Sea cucumbers are processed into bêche-de-mer by both domestic and industrial level processors following the similar steps of cleaning, evisceration, first boiling, salting, second boiling and drying. However, domestically processed bêche-de-mer always reported a higher percentage of moisture, crude ash, crude fat and lower percentage of crude protein than industrially processed products. Although processing resulted in a significant reduction of total SFA and MUFA in fresh individuals of most of these species, total PUFA increased significantly in processed individuals excluding Bohadschia species. Palmitic acid was found to be the most dominant fatty acid in all these species followed by eicosapentaenoic acid, which showed a significant increase in processed products, except Bohadschia sp. 1. Total MUFA were higher than total SFA in all sea cucumber species having exceptions in Bohadchia sp.1 and fresh S. chloronotus. These findings will make a significant contribution to fill the gaps in existing information as no any previous information is available for species like H. spinifera and S. chloronotus.
Product Description
Supelco
Supelco 37 Component FAME Mix, certified reference material, TraceCERT®, in dichloromethane (varied conc.), ampule of 1 mL