  • Search Results
  • A novel circular RNA, hsa_circ_0046701, promotes carcinogenesis by increasing the expression of miR-142-3p target ITGB8 in glioma.
A novel circular RNA, hsa_circ_0046701, promotes carcinogenesis by increasing the expression of miR-142-3p target ITGB8 in glioma.
Biochemical and biophysical research communications (2018-01-18)
Guobin Li, Haiying Yang, Kun Han, Dezhang Zhu, Peng Lun, Yan Zhao
ABSTRACT
In the present study, we identified a novel circular RNA (circRNA), hsa_circ_0046701, in glioma cells. We measured the expression of hsa_circ_0046701 using qRT-PCR in glioma tissues and cell lines, and explored its functions using 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide, colony formation, and Transwell assays. Luciferase reporter assays were performed to validate the correlation between microRNA miR-142-3p and hsa_circ_0046701 or integrin subunit beta 8 (ITGB8). The results showed that hsa_circ_0046701 was significantly upregulated in glioma tissues and cell lines, and knockdown of hsa_circ_0046701 inhibited cell proliferation and invasion. Luciferase reporter assays indicated that hsa_circ_0046701 functions as a sponge for miR-142-3p and regulates the expression of ITGB8. Subsequently, functional assays revealed that silencing of hsa_circ_0046701 could upregulate miR-142-3p, resulting in downregulation of ITGB8. The results demonstrated that the hsa_circ_0046701/miR-142-3p/ITGB8 axis might play critical regulatory roles in the pathogenesis and development of glioma.
Sigma-Aldrich
GeneJuice® Transfection Reagent, Non-lipid based chemical transfection reagent optimized for maximum transfection efficiency, ease-of-use, and minimal cytotoxicity on a wide variety of mammallian cells.
Sigma-Aldrich
Fetal Bovine Serum, USA origin, Heat Inactivated, sterile-filtered, suitable for cell culture, suitable for insect cell culture, suitable for hybridoma