  • Search Results
  • A microfluidic biochip platform for electrical quantification of proteins.
A microfluidic biochip platform for electrical quantification of proteins.
Lab on a chip (2018-04-18)
Enrique Valera, Jacob Berger, Umer Hassan, Tanmay Ghonge, Julia Liu, Michael Rappleye, Jackson Winter, Daniel Abboud, Zeeshan Haidry, Ryan Healey, Na-Teng Hung, Nathaniel Leung, Naif Mansury, Alexander Hasnain, Christine Lannon, Zachary Price, Karen White, Rashid Bashir
ABSTRACT
Sepsis, an adverse auto-immune response to an infection often causing life-threatening complications, results in the highest mortality and treatment cost of any illness in US hospitals. Several immune biomarker levels, including Interleukin 6 (IL-6), have shown a high correlation to the onset and progression of sepsis. Currently, no technology diagnoses and stratifies sepsis progression using biomarker levels. This paper reports a microfluidic biochip platform to detect proteins in undiluted human plasma samples. The device uses a differential enumeration platform that integrates Coulter counting principles, antigen specific capture chambers, and micro size bead based immunodetection to quantify cytokines. This microfluidic biochip was validated as a potential point of care technology by quantifying IL-6 from plasma samples (n = 29) with good correlation (R2 = 0.81) and agreement (Bland-Altman) compared to controls. In combination with previous applications, this point of care platform can potentially detect cell and protein biomarkers simultaneously for sepsis stratification.
Sigma-Aldrich
Streptavidin from Streptomyces avidinii, essentially salt-free, lyophilized powder, ≥13 units/mg protein
Sigma-Aldrich
Bovine Serum Albumin, heat shock fraction, protease free, essentially globulin free, pH 7, ≥98%
Sigma-Aldrich
N-Hydroxysulfosuccinimide sodium salt, ≥98% (HPLC)