  • Search Results
  • Kallikrein-8 inhibition attenuates Alzheimer's disease pathology in mice.
Kallikrein-8 inhibition attenuates Alzheimer's disease pathology in mice.
Alzheimer's & dementia : the journal of the Alzheimer's Association (2016-06-22)
Arne Herring, Yvonne Münster, Tamer Akkaya, Sahar Moghaddam, Katharina Deinsberger, Jakob Meyer, Julia Zahel, Eduardo Sanchez-Mendoza, Yachao Wang, Dirk M Hermann, Thomas Arzberger, Sarah Teuber-Hanselmann, Kathy Keyvani
ABSTRACT
Memory loss and increased anxiety are clinical hallmarks of Alzheimer's disease (AD). Kallikrein-8 is a protease implicated in memory acquisition and anxiety, and its mRNA is known to be up-regulated in AD-affected human hippocampus. Therefore, an involvement of Kallikrein-8 in Alzheimer's pathogenesis is conceivable but remains to be proved. We determined the cerebral expression of Kallikrein-8 mRNA and protein during the course of AD in patients and in transgenic mice and tested the impact of Kallikrein-8 inhibition on AD-related pathology in mice and in primary glial cells. Kallikrein-8 mRNA and protein were up-regulated in both species at incipient stages of AD. Kallikrein-8 inhibition impeded amyloidogenic amyloid-precursor-protein processing, facilitated amyloid β (Aβ) clearance across the blood-brain-barrier, boosted autophagy, reduced Aβ load and tau pathology, enhanced neuroplasticity, reversed molecular signatures of anxiety, and ultimately improved memory and reduced fear. Kallikrein-8 is a promising new therapeutic target against AD.
Sigma-Aldrich
Anti-ATG5 (N-terminal) antibody produced in rabbit, affinity isolated antibody, PBS solution
Sigma-Aldrich
Anti-Laminin antibody produced in rabbit, 0.5 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Rat IgG (whole molecule)−Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
EphB2/Fc Chimera from mouse, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Sigma-Aldrich
Anti-LRP, Light Chain Mouse mAb (5A6), liquid, clone 5A6, Calbiochem®
Sigma-Aldrich
Anti-Tau Antibody, a.a. 210-241, clone Tau-5, ascites fluid, clone Tau-5, Chemicon®