US
EN
Applications
Products
Services
Support
Home
Search Results
Water, Deionized Distilled, ASTM Type II
(2)
Water, Deionized Distilled, ASTM Type II
Synonyms:
Water
Linear Formula:
H
2
O
CAS Number:
7732-18-5
Molecular Weight:
18.02
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
6442
Water, Deionized Distilled, ASTM Type II, HARLECO
®
Purified water for laboratory use
Expand
View Pricing
EM3234
Water, Deionized Distilled, ASTM Type II, Distilled, ASTM Type II
Expand
View Pricing