MilliporeSigma
Triton™ X-100 Detergent, Molecular Biology Grade Non-ionic detergent and emulsifier. Has an optimal pH range of 6.0-8.0.

Triton X-100 Detergent, Molecular Biology Grade

Triton™ X-100 Detergent, Molecular Biology Grade Non-ionic detergent and emulsifier. Has an optimal pH range of 6.0-8.0.
Synonyms:
Triton X-100, t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether
Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Triton X-100 Detergent, Molecular Biology Grade, Non-ionic detergent and emulsifier. Has an optimal pH range of 6.0-8.0.