Triton™ X-100 for gas chromatography

Triton X-100

Triton™ X-100 for gas chromatography
Synonyms:
t-Octylphenoxypolyethoxyethanol, Polyethylene glycol tert-octylphenyl ether
Linear Formula:
t-Oct-C6H4-(OCH2CH2)xOH, x= 9-10
CAS Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Triton X-100, BioXtra
Triton X-100, laboratory grade
Triton X-100, for molecular biology
Triton X-100, non-ionic, aqueous solution
Triton X-100, EMPROVE® EXPERT Ph Eur
Triton X-100, for analysis
Triton X-100, peroxide- and carbonyl-free
Triton X-100, for gas chromatography