Apocynin
Synonym(s):
4′-Hydroxy-3′-methoxyacetophenone, Acetovanillone, Apocynin
Linear Formula:
HOC6H3(OCH3)COCH3
CAS Number:
Molecular Weight:
166.17
Products
Shipping
Boiling Point (°C)
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing