US
EN
Applications
Products
Services
Documents
Support
Home
Search Results
Sodium hydroxide solution min. 10% (1.11)
(1)
Sodium hydroxide solution min. 10% (1.11)
Synonyms:
Sodium hydroxide solution
Linear Formula:
NaOH
CAS Number:
1310-73-2
Molecular Weight:
40.00
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Showing 1-1 of 1
Page 1 of 1
Page 1 of 1
Compare
Product Number
Product Description
Pricing
1.05588
Sodium hydroxide solution min. 10% (1.11),
for analysis EMSURE
®
Expand
View Pricing
Page 1 of 1
Page 1 of 1