MilliporeSigma
Sodium hydroxide solution 21% for analysis EMSURE®

Sodium hydroxide solution 21%

Sodium hydroxide solution 21% for analysis EMSURE®
Synonyms:
Sodium hydroxide solution
Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Sodium hydroxide solution 21%, for analysis EMSURE®