Products
US
EN
Products
Applications
Products
Services
Support
Home
Search Results
Sodium hydroxide solution 10 mol/L
(1)
Sodium hydroxide solution 10 mol/L
Synonyms:
Sodium hydroxide solution
Linear Formula:
NaOH
CAS Number:
1310-73-2
Molecular Weight:
40.00
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
4.80648
Sodium hydroxide solution 10 mol/L, EMPROVE
®
EXPERT
Expand
View Pricing