MilliporeSigma
Sodium hydroxide 1.000 l for the preparation of dilute volumetric solutions or for direct use c(NaOH) = 5 mol/l (5 N) Combi-Titrisol

Sodium hydroxide 1.000 l

Sodium hydroxide 1.000 l for the preparation of dilute volumetric solutions or for direct use c(NaOH) = 5 mol/l (5 N) Combi-Titrisol
Synonyms:
Sodium hydroxide solution
Linear Formula:
NaOH
CAS Number:
Molecular Weight:
40.00

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Sodium hydroxide 1.000 l, for the preparation of dilute volumetric solutions or for direct use c(NaOH) = 5 mol/l (5 N) Combi-Titrisol