MilliporeSigma
Sodium hydrogen selenite purum p.a., ≥96.0% (RT)

Sodium hydrogen selenite

Sodium hydrogen selenite purum p.a., ≥96.0% (RT)
Synonyms:
Sodium hydroselenite, Sodium biselenite, Sodium hydrogen selenite
Linear Formula:
NaHSeO3
CAS Number:
Molecular Weight:
150.96

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Sodium hydrogen selenite, purum p.a., ≥96.0% (RT)
Sodium hydrogen selenite, suitable for microbiology