MilliporeSigma
Rifampin United States Pharmacopeia (USP) Reference Standard

Rifampin

Rifampin United States Pharmacopeia (USP) Reference Standard
Synonyms:
Rifampicin, 3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifampin, Rifamycin AMP
Empirical Formula (Hill Notation):
C43H58N4O12
CAS Number:
Molecular Weight:
822.94

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Rifampin, Pharmaceutical Secondary Standard; Certified Reference Material
Rifampin, United States Pharmacopeia (USP) Reference Standard