MilliporeSigma
Rifampicin European Pharmacopoeia (EP) Reference Standard

Rifampicin

Rifampicin European Pharmacopoeia (EP) Reference Standard
Synonyms:
3-(4-Methylpiperazinyliminomethyl)rifamycin SV, Rifampin, Rifamycin AMP
Empirical Formula (Hill Notation):
C43H58N4O12
CAS Number:
Molecular Weight:
822.94

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Rifampicin, ≥97% (HPLC), powder
Rifampicin, suitable for plant cell culture, BioReagent, ≥97% (HPLC), crystalline
Rifampicin, European Pharmacopoeia (EP) Reference Standard