MilliporeSigma
OmniPur® TRIS

OmniPur® TRIS

OmniPur® TRIS
Synonyms:
Tris base, Trometamol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, THAM, Tris(hydroxymethyl)aminomethane
Linear Formula:
NH2C(CH2OH)3
CAS Number:
Molecular Weight:
121.14

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
OmniPur® TRIS