US
EN
Applications
Products
Services
Support
Home
Search Results
Nitric acid concentrate
(1)
Nitric acid concentrate
Synonyms:
Nitric acid solution
Empirical Formula (Hill Notation):
HNO
3
CAS Number:
7697-37-2
Molecular Weight:
63.01
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
16355
Nitric acid concentrate, 0.1 M HNO
3
in water (0.1N), eluent concentrate for IC
Expand
View Pricing