US
EN
Applications
Products
Services
Support
Home
Search Results
Magnesium Sulfate, Anhydrous
(2)
Magnesium Sulfate, Anhydrous
Synonyms:
Magnesium sulfate anhydrous, Magnesium Sulfate, Anhydrous
Empirical Formula (Hill Notation):
MgO
4
S
CAS Number:
7487-88-9
Molecular Weight:
120.37
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
1.06067
Magnesium Sulfate, Anhydrous, for analysis EMSURE
®
Expand
View Pricing
MX0075
Magnesium Sulfate, Anhydrous, powder Meets Reagent Specifications for testing USP/NF monographs GR
Expand
View Pricing