MilliporeSigma
Glycine, Molecular Biology Grade - CAS 56-40-6 - Calbiochem Absorbance (260 nm and 280 nm): ≤0.01.

Glycine, Molecular Biology Grade - CAS 56-40-6 - Calbiochem

Glycine, Molecular Biology Grade - CAS 56-40-6 - Calbiochem Absorbance (260 nm and 280 nm): ≤0.01.
Synonyms:
Glycine, Aminoacetic acid, Aminoethanoic acid, Glycocoll
Linear Formula:
NH2CH2COOH
CAS Number:
Molecular Weight:
75.07

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Glycine, Molecular Biology Grade - CAS 56-40-6 - Calbiochem, Absorbance (260 nm and 280 nm): ≤0.01.