MilliporeSigma
Ethyl Alcohol Non UV, denatured OmniSolv®

Ethyl Alcohol

Ethyl Alcohol Non UV, denatured OmniSolv®
Synonyms:
Alcohol
Empirical Formula (Hill Notation):
C2H6O
CAS Number:
Molecular Weight:
46.07

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Ethyl Alcohol, 95% denatured
Ethyl Alcohol, anhydrous, denatured
Ethyl Alcohol, 95% denatured, special
Ethyl Alcohol, Non UV, denatured OmniSolv®