Products
US
EN
Products
Applications
Products
Services
Support
Home
Search Results
Ethanol Fixative 80% v/v
(1)
Ethanol Fixative 80% v/v
Synonyms:
Ethanol solution, Ethyl alcohol
Linear Formula:
C
2
H
5
OH
CAS Number:
64-17-5
Molecular Weight:
46.07
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
2858
Ethanol Fixative 80% v/v, suitable for fixing solution (blood films)
Expand
View Pricing