US
EN
Applications
Products
Services
Support
Home
Search Results
Dimethylformamide
(2)
Dimethylformamide
Synonyms:
N,N
-Dimethylformamide, DMF, NSC 5356
Linear Formula:
HCON(CH
3
)
2
CAS Number:
68-12-2
Molecular Weight:
73.09
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
PHR1553
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing
RTC000098
Dimethylformamide, Pharmaceutical Secondary Standard; Certified Reference Material
Expand
View Pricing