US
EN
Applications
Products
Services
Support
Home
Search Results
Dextran 4 for calibration
(1)
Dextran 4 for calibration
Synonyms:
Dextran
Linear Formula:
[C
6
H
10
O
5
]
n
CAS Number:
9004-54-0
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
D0733000
Dextran 4 for calibration, European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing