US
EN
Applications
Products
Services
Support
Home
Search Results
Dextran 1
(2)
Dextran 1
Synonyms:
Dextran
Linear Formula:
[C
6
H
10
O
5
]
n
CAS Number:
9004-54-0
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
D0731005
Dextran 1, European Pharmacopoeia (EP) Reference Standard
Expand
View Pricing
1179628
Dextran 1, United States Pharmacopeia (USP) Reference Standard
Expand
View Pricing