US
EN
Applications
Products
Services
Support
Home
Search Results
Cholesteryl chloride
(2)
Cholesteryl chloride
Synonyms:
3β-Chloro-5-cholestene, Cholesteryl chloride
Empirical Formula (Hill Notation):
C
27
H
45
Cl
CAS Number:
910-31-6
Molecular Weight:
405.10
Product Comparison Guide
Use the product attributes below to configure the comparison table. (Select up to 3 total.)
Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Select Attributes
Product Number
Product Description
Pricing
C76604
Cholesteryl chloride, 97%
Expand
View Pricing
8.41733
Cholesteryl chloride, for synthesis
Expand
View Pricing