MilliporeSigma
Carbon disulfide solution 5.0 M in hexane

Carbon disulfide solution

Carbon disulfide solution 5.0 M in hexane
Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Carbon disulfide solution, certified reference material, 5000 μg/mL in methanol
Carbon disulfide solution, 5 M in THF
Carbon disulfide solution, 5 M in ethanol
Carbon disulfide solution, 5.0 M in hexane