MilliporeSigma
Carbon disulfide ReagentPlus®, purified by redistillation, ≥99.9%

Carbon Disulfide

Carbon disulfide ReagentPlus®, purified by redistillation, ≥99.9%
Synonyms:
Carbon disulfide, Carbon Disulfide
Empirical Formula (Hill Notation):
CS2
CAS Number:
Molecular Weight:
76.14

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Carbon disulfide, anhydrous, ≥99%
Carbon disulfide, ACS reagent, ≥99.9%
Carbon disulfide, suitable for HPLC, ≥99.9%
Carbon disulfide, ACS reagent, ≥99.9%
Carbon disulfide, ReagentPlus®, low benzene, ≥99.9%
Carbon disulfide, for analysis EMSURE® ACS,Reag. Ph Eur
Carbon disulfide, suitable for IR spectroscopy, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (GC)
Carbon Disulfide, low benzene OmniSolv®
Carbon Disulfide, Meets Reagent Specifications for testing USP/NF monographs OmniSolv®
Carbon Disulfide, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS
Carbon disulfide, EMPLURA®
Carbon disulfide, for spectroscopy Uvasol®
Carbon disulfide, ReagentPlus®, purified by redistillation, ≥99.9%
Carbon disulfide, Emplura®
Carbon Disulfide, Pharmaceutical Secondary Standard; Certified Reference Material