MilliporeSigma
Carbon dioxide (99.8%) cylinder of 14 L, analytical standard

Carbon dioxide (99.8%)

Carbon dioxide (99.8%) cylinder of 14 L, analytical standard
Empirical Formula (Hill Notation):
CO2
Molecular Weight:
44.01
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Carbon dioxide (99.8%), cylinder of 48 L, analytical standard
Carbon dioxide (99.8%), cylinder of 14 L, analytical standard