Benzo[ghi]perylene solution certified reference material, 200 μg/mL in dichloromethane

Benzo[ghi]perylene solution

Benzo[ghi]perylene solution certified reference material, 200 μg/mL in dichloromethane
Empirical Formula (Hill Notation):
C22H12
CAS Number:
Molecular Weight:
276.33
EC Number:

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Benzo[ghi]perylene solution, certified reference material, 200 μg/mL in dichloromethane