MilliporeSigma
Bafilomycin A1 from Streptomyces griseus ≥90% (HPLC)

Bafilomycin A1 from Streptomyces griseus

Bafilomycin A1 from Streptomyces griseus ≥90% (HPLC)
Empirical Formula (Hill Notation):
C35H58O9
CAS Number:
Molecular Weight:
622.83

Product Comparison Guide

Use the product attributes below to configure the comparison table. (Select up to 3 total.)

Select Attribute
Select Attribute
Select Attribute
Sort by: Default
Product Number
Product Description
Pricing
Bafilomycin A1 from Streptomyces griseus, ≥90% (HPLC)